Learn More
Kinesis™ PEEK Chromatography Tubing
An excellent replacement for stainless steel tubing
Brand:  Kinesis™ 02006-33
 View more versions of this product
Product Code. 13009902
  • 431.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
  • Biocompatible, inert and easy to cut
  • Great for high-pressure applications
  • Wide choice of outer and inner diameters
  • Many sizes color-coded for easier inner diameter identification
Specifications
Specifications
Tubing
High-Pressure Applications
50 ft.
7000 psi (483 bar)
Yellow
0.007 in.
PEEK