Learn More
Kinesis™ Plastic Tubing Cutter
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
15.25 €
Description
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
- Built-in guide holes for both 1/16 and 1/8 in. OD tubing
- Replacement blades included and sold separately
Offers durability, reliability and ease
Specifications
Upchurch™ standard polymer tubing cutter |