missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Quick Connect Adapter

Convenience of quick connect and disconnect of a luer connection

23.95 € - 42.30 €

Specifications

Diameter (English) Inner 0.05 in.
View More Specs

Products 2
Product Code Brand Color Type For Use With (Equipment) Temperature (Metric) Max Price Quantity & Availability  
Product Code Brand Color Type For Use With (Equipment) Temperature (Metric) Max Price Quantity & Availability  
12937759
View Documents
Kinesis™
Tan Female Luer Adapter Female Luer x Female 10-32 Coned
42.30 €
Each
In Stock
to see stock.
Add to basket
 
12967769
View Documents
Kinesis™
Red Male Luer Adapter Male Luer Lock x Female 1/4-28 Flat Bottom 40°C
23.95 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Convenience of quick connect and disconnect of a luer connection

Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
Specifications

Specifications

0.05 in.
Videos
SDS
Documents

Documents

Product Certifications