Learn More
Kinesis™ Threaded Plugs and Caps
Plugs to close off unused ports in valves and multi-port connectors
- Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Brand: Kinesis™ P-553
This item is not returnable.
View return policy
Description
Plugs to close off unused ports in valves and multi-port connectors
- Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Specifications
Plug | |
White | |
6-32 Coned | |
Delrin |