missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific Upright Freezer Rack, Stainless Steel
GREENER_CHOICE

The Upright Freezer Rack safely stores samples in your freezer. Constructed of corrosion-resistant stainless steel racks that are compatible with major brands of upright freezers.

Brand:  Heathrow Scientific HS2862KB

 View more versions of this product

Product Code. 16370766

  • 239.00 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Upright Freezer Rack
  • Rack has handle for easy transfer
  • Robust mounting brackets assure handles stay securely in place for easy use
  • Locking Rod Keeps tube storage boxes safely in place
Specifications

Specifications

Upright Freezer Rack
Silver
Stainless Steel
Tube Boxes
16
Box
22 in.
5.5 in.
12.8 in.
4 x 4
Boxes (3 in.)
Non-sterile
HS2862KB
1 Ea.
55.6 cm
14.1 cm
32.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.