missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Upright ULT Freezer Drawer Racks for Centrifuge Tubes

Fisherbrand inventory control racks are ideal for all cold storage application. Provide an ideal alternative for storing large quantities of samples in 15mL or 50mL centrifuges tubes.

Brand:  Fisherbrand™ UFD-LT50-2

 View more versions of this product

Product Code. 15709559

  • 237.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • For 15mL or 50mL centrifuges tubes
  • 2 drawers, holds between 78 and 160 tubes
  • Made of high quality stainless steel
Specifications

Specifications

Drawer Rack
Stainless Steel
Upright ULT Freezer
Conical Tubes (78 x 50 mL)
50ml conical tubes
559 x 252 x 140mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers