Learn More
Fisherbrand™ Upright ULT Freezer Racks
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of upright ULT freezers.
Brand:  Fisherbrand™ UF-332
 View more versions of this product
Product Code. 15719539
  • 102.00 € / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Side opening with handles on the front for easy box retrieval.
Specifications
Specifications
Side Access Freezer Rack
Stainless Steel
Upright ULT Freezer
9 (2 in.) boxes
Standard 2 in. Cryoboxes
415 x 140 x 168 mm