Learn More
Fisherbrand™ Upright ULT Freezer Racks
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of upright ULT freezers.
Brand:  Fisherbrand™ UF-42-375
 View more versions of this product
Product Code. 15759539
  • 109.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Side opening with handles on the front for easy box retrieval.
Specifications
Specifications
Side Access Freezer Rack
Stainless Steel
Upright ULT Freezer
8 (100mm) boxes
8 (100mm) boxes
550 x 140 x 205 mm