missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
33.40 € - 324.00 €
Chemical Identifiers
CAS | 57-13-6 |
---|---|
Molecular Formula | CH4N2O |
Molecular Weight (g/mol) | 60.06 |
InChI Key | XSQUKJJJFZCRTK-UHFFFAOYSA-N |
Synonym | carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate |
PubChem CID | 1176 |
ChEBI | CHEBI:48376 |
IUPAC Name | urea |
SMILES | C(=O)(N)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10123430
|
Thermo Scientific Acros
140750010 |
1 kg | Plastic bottle |
33.40 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10665572
|
Thermo Scientific Acros
140750050 |
5 kg | Plastic drum |
92.80 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11906921
|
Thermo Scientific Acros
140750250 |
25 kg | Plastic bag in resealable cardbox |
324.00 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemical Identifiers
57-13-6 | |
60.06 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
CHEBI:48376 | |
C(=O)(N)N |
CH4N2O | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
1176 | |
urea |
Specifications
57-13-6 | |
White | |
Authentic | |
CH4N2O | |
03,42 | |
15,10052 | |
Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol,benzene,concentrated hcl,practically insoluble in chloroform and ether,soluble in methanol: 16.6g/100 mL,soluble in ethanol: 5g/100 mL | |
C(=O)(N)N | |
60.06 | |
CHEBI:48376 | |
99.5% | |
0.01% max | |
Urea |
131.0°C to 135.0°C | |
0.01% max | |
99.5% | |
H2NCONH2 | |
01,1262; 02,455 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
urea | |
1176 | |
60.06 | |
Analysis | |
Fine Crystals |
Safety and Handling
missing translation for 'einecsNumber' : 200-315-5
RUO – Research Use Only