missing translation for 'onlineSavingsMsg'
Learn More
Urea, 99%, Thermo Scientific Chemicals Product Code.: 10200001

Urea, 99%, Thermo Scientific Chemicals

Product Code. 10200001
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10200001

Brand: Thermo Scientific Acros

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Decomposition Information >132°C
Melting Point 131.0°C to 135.0°C
Color White
Ignition Residue 0.01% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Molecular Formula CH4N2O
Linear Formula H2NCONH2
Quantity 500 g
Beilstein 03, 42
Fieser 01,1262; 02,455
Merck Index 15, 10052
Solubility Information 0.01% max. Insoluble Matter
Molecular Weight (g/mol) 60.056
Formula Weight 60.06
Vapor Pressure 1.25mmHg at 25°C
Percent Purity 99%
Grade ACS Reagent
Physical Form Fine Crystals
Chemical Name or Material Urea
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0431020 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number424585000,424581000,424580100,424580025
Lot NumberA0463579 Certificate TypeCertificate of Analysis Date07/06/2024 Catalog Number424585000,424581000,424580100,424580025
Lot NumberB0150155A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number424585000,424581000,424580100,424580025
Lot NumberB0148758 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number424585000,424581000,424580100,424580025
Lot NumberB0147922 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number424585000,424581000,424580100,424580025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Urea, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.