missing translation for 'onlineSavingsMsg'
Learn More

Urea, Honeywell Fluka™
BIOPHARMA

Product Code. 16488313
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES NC(N)=O

Specifications

Physical Form Crystalline Granules
Quantity 1 kg
Percent Purity ≥99.5%
Loss on Drying Max. 1.0 %
Packaging Plastic Bottle
Appearance Complying
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'