missing translation for 'onlineSavingsMsg'
Learn More

Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2, Thermo Scientific Chemicals

Product Code. 11924381
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-43-6, 7722-88-5
Molecular Formula CH6N2O3
Molecular Weight (g/mol) 94.07
InChI Key AQLJVWUFPCUVLO-UHFFFAOYSA-N
Synonym urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid
PubChem CID 31294
ChEBI CHEBI:75178
IUPAC Name hydrogen peroxide;urea
SMILES C(=O)(N)N.OO

Specifications

Melting Point 75.0°C to 85.0°C
Color White
Packaging Plastic drum
Linear Formula CO(NH2)2·H2O2
Quantity 5 kg
Fieser 16,379; 17,401
Merck Index 15,1784
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: alcohol and ether decomposes the product to h2o2, and urea, soluble in ethylene glycol
Stabilizer 1% sodium pyrophosphate
Formula Weight 94.07
Percent Purity ≥33.5% (H2O2)
Physical Form Tablets
Chemical Name or Material Urea hydrogen peroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Urea hydrogen peroxide (1 g tablets), stabilized, contains 35 wt% H2O2
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'