missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, puriss. p.a., ACS reagent, ≥99.5%, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
164.71 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
2.5 kg
Specifications
CAS | 57-13-6 |
CAS Max % | 1 |
Color | White |
Density | 1.32 g/cm3 (20°C) |
pH | 9 to 10 |
Molecular Formula | CH4N2O |
Quantity | 2.5 kg |
InChI Key | XSQUKJJJFZCRTK-UHFFFAOYSA-N |
SMILES | NC(N)=O |
IUPAC Name | urea |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'