Learn More
Uricase, Candida utilis, Thermo Scientific Chemicals
Description
Uricase is very important for the determination of uric acid in biological fluids. Uricase is an important medical enzyme which can be used to determine urate in clinical analysis. The enzyme Urate oxidase (UO) catalyzes the oxidation of uric acid to 5-hydroxyisourate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 9002-12-4 |
Packaging | Plastic bottle |
Molecular Formula | (C3H4O2)n |
MDL Number | MFCD00084394 |
Quantity | 100 U |
Synonym | acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer |
Solubility Information | Hardly soluble in water,slightly soluble in buffer alkaline solution. |
InChI Key | NNMHYFLPFNGQFZ-UHFFFAOYSA-M |
SMILES | OC(=O)C(-*)C-* |
IUPAC Name | prop-2-enoic acid |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P05L034 | Certificate of Analysis | 10/03/2025 | J60875.!R |
N06L010 | Certificate of Analysis | 10/02/2025 | J60875.!R |
Z12K028 | Certificate of Analysis | 23/01/2025 | J60875.!R |
Y21K013 | Certificate of Analysis | 26/11/2024 | J60875.!R |
X18K027 | Certificate of Analysis | 05/11/2024 | J60875.!R |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.