Learn More
Ursonic acid, MedChemExpress
MedChemExpress Ursolic acid, a naturally occurring triterpenoid, induces the apoptosis of human cancer cells through multiple signaling pathways.
66.15 € - 79.65 €
Specifications
Chemical Name or Material | Ursonic acid |
---|---|
CAS | 6246-46-4 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C30H46O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16499228
|
Medchem Express
HY-N1486 |
10 mg |
79.65 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16431648
|
Medchem Express
HY-N1486 |
5 mg |
66.15 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ursolic acid, a naturally occurring triterpenoid, induces the apoptosis of human cancer cells through multiple signaling pathways.Specifications
Ursonic acid | |
White | |
C30H46O3 | |
DMSO : 50 mg/mL (109.97 mM; ultrasonic and warming and heat to 80°C) ∣H2O : 1 mg/mL (2.20 mM; ultrasonic and warming and heat to 80°C) | |
454.68 | |
98.15% | |
Solid |
6246-46-4 | |
Research | |
3-Ketoursolic acid | |
CC1(C)C(CC[C@]2(C)[C@@]3([H])CC=C4[C@]5([H])[C@@H](C)[C@H](C)CC[C@@](C(O)=O)5CC[C@](C)4[C@@](C)3CC[C@@]12[H])=O | |
454.68 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.