Learn More
USB Device Interface Kit, for OHAUS™ Scout STX and SKX Precision Portable Balances
A unique solution to the problem of connecting a balance to a computer using a USB. The USB Interface kit includes a CD with the software drivers to create the required virtual port on the computer.
Brand:  Ohaus 30268984
Product Code. 15509641
  • 91.25 € / Each
Explore more special offers
Description
Description
System Requirements
  • PC running Windows 98™, Windows 98SE™, Windows ME™, Windows 2000™, Windows XP™ or Windows 7™
  • Be forward compatible with iMac 10.7
  • Available USB port
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Interface Cables and Kits