Learn More
USB Host Interface Kit, for OHAUS™ Scout STX and SKX Precision Portable Balances
Helps to export or import the menu settings and application mode setting parameter.
Brand: Ohaus 30268983
129.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Weighing results can also be saved to the USB flash drive.
Specifications
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) | |
Interface Cables and Kits |
Ohaus Scout precision portable balances SKX series
File size: 165.4 kB
Product Content Correction
Product Title
USB Host Interface Kit, for OHAUS™ Scout STX and SKX Precision Portable Balances > USB Host Interface Kit
Spot an opportunity for improvement?Share a Content Correction