Learn More
Memmert™ USB Stick with Documentation Software AtmoCONTROL and Operation Manual for SF75
USB stick accessory for Memmert Sterilizers.
Brand:  Memmert™ SF75B33172
Product Code. 17528395
  • 196.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • USB stick with documentation software AtmoCONTROL and operation manual for products with SingleDISPLAY (the standard equipment of appliances with TwinDISPLAY includes one USB stick with AtmoCONTROL)
  • When reordering please specify serial number
Specifications
Specifications
SF75 Sterilizer
USB Stick
Documentation Software AtmoCONTROL and Operation Manual