missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP33 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12060-2-AP-150UL
This item is not returnable.
View return policy
Description
DNA-directed RNA polymerase I subunit RPA43 or Twist neighbor protein (Twistnb)belongs to the eukaryotic RPA43 RNA polymerase subunit family. It catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates.Twistnb is the component of RNA polymerase I which synthesizes ribosomal RNA precursors. It is widely expressed in all fetal and adult tissues, with highest expression in fetal lung, liver, and kidney, and low expression in all adult tissues. Twist genes are essential for embryonic development and are conserved from jellyfish to human.Specifications
USP33 | |
Polyclonal | |
Unconjugated | |
USP33 | |
Deubiquitinating enzyme 33, hVDU1, KIAA1097, Ubiquitin thiolesterase 33, USP33, VDU1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170822, 23032, 310960 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8R5K2, Q8TEY7 | |
Usp33 | |
USP33 Fusion Protein Ag2695 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction