missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP49 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18066-1-AP-150UL
This item is not returnable.
View return policy
Description
Specifically deubiquitinates histone H2B at ′Lys-120′ (H2BK120Ub). H2BK120Ub is a specific tag for epigenetic transcriptional activation and acts as a regulator of mRNA splicing. Deubiquitination is required for efficient cotranscriptional splicing of a large set of exons.Specifications
USP49 | |
Polyclonal | |
Unconjugated | |
USP49 | |
Deubiquitinating enzyme 49, Ubiquitin thiolesterase 49, USP49 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
224836, 25862, 316211 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P9L4, Q70CQ1 | |
Usp49 | |
USP49 Fusion Protein Ag12697 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |