Learn More
Cole-Parmer™ Economical Utility Gauge
The Economical Utility Gauge is a high-quality, low cost gauge designed for industrial processes.
Brand:  Cole-Parmer™ 68355-30
 View more versions of this product
Product Code. 11799829
  • 9.30 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Utility gauges are designed to replace more expensive digital or commercial gauges
Specifications
Specifications
CE Compliance
0.1 in. NPT(M), Lower
3-2-3% Full-scale
0 to 300 psi
High-quality, low cost gauges designed for industrial processes
3.17 mm NPT(M), Lower
Analog
Utility Pressure Gauge
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Economical Utility Gauge > Utility Gauge; Range: 0 to 300 psi