missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Utility Trays for Shaker Platforms

Use these utility trays with 0.5 in. rubber mats with Thermo Scientific™ shakers.

Brand:  Thermo Scientific™ 75004123

 View more versions of this product

Product Code. 17831905

  • 268.34 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use these Thermo Scientific™ Utility trays for Shaker Platforms with 0.5 in. rubber mats.

Mix and match Thermo Scientific™ Solaris shaker Universal clamps to suit individual needs in the laboratory.  Compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers, MaxQ6000 Shakers.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

353 x 272 mm
11 x 14 in. and 12 x 14 in.
Utility Tray for Platforms
Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.