missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benchmark Scientific™ UV-Clave Ultraviolet Chamber For Research Use Only
Short-wave UV light is an effective method for the disinfection of surface DNA, RNA and microorganisms. With the UV-Clave chamber, highly focused UV irradiation is used to disinfect tools, vessels and other research items, eliminating up to 99% of surface contamination.
Brand: Benchmark Scientific™ B1450-E
Description
- Improves quality of PCR research by reducing potential contaminants
- Multiple lamps and reflective chamber walls provide true 360° irradiation
- Proprietary UVC transparent shelf, eliminates potential for missed coverage (blind spots)
- Inactivates DNA, RNA, viruses and bacteria on PCR vessels
- Adjustable cycle
- Lights turn off then the door is opened to protect users
Specifications
1047 cu. in. | |
25.6 cm | |
10.1 in. | |
Sterilization of PCR Tools | |
29.2 cm | |
11 lb. | |
5 kg | |
12.2 in | |
230 V | |
21.6 x 25.6 x 31.8 cm |
1758 cu. cm | |
8.5 in. | |
21.6 cm | |
11.5 in. | |
UV Sanitizer Cabinet | |
Digital | |
31.8 cm | |
2 Stainless Steel Wire Shelves, 1 UV-C Transparent Shelf | |
Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction