Learn More
Cytiva UV Flow Cell ÅKTApilot
UV Flow cell ÅKTApilot can also be used with spectrohotometer model Uvis-920
Brand:  Cytiva 11-0008-50
Product Code. 11390192
  • 2813.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Monitor UVis-920 is a high-precision, multipurpose, standalone UV monitor. The use of different filters allows the monitor to cover wavelengths from 200nm to 700nm, without changing the lamp. Pre-assembled filters with holders are placed with a push-button mechanism. Other wavelengths can be achieved by using an appropriate filter assembled in an empty filter holder. The fibre optics, together with flow cell configurations, cover a wide flow range for a variety of applications While providing excellent flow cell accessibility. This allows columns to be placed close to the flow cell, which reduces band broadening and gives a high signal-to-noise ratio. This feature, together with a low noise and a low thermal drift, will allow a sensitive, stable and reliable UV/Vis absorbance detection. UV Flow cell ÅKTApilot can also be used with spectrohotometer model Uvis-920.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UVis-920
UV Flow Cell ÄKTApilot
Optical fiber
1 Each