missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vilber Lourmat™ UV Lamp
Single wavelength UV lamp with filter that is an ideal solution for research and enumeration of E. coli.
271.00 € - 969.00 €
Product Code | Brand | Lighting Intensity | Wattage | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lighting Intensity | Wattage | Price | Quantity & Availability | ||||
11776518
|
Vilber Lourmat™
311100451 |
265/350 mW/cm2 | 8 W | N/A | |||||
11820430
|
Vilber Lourmat™
311100411 |
340 mW/cm2 | 4 W | N/A | |||||
11860380
|
Vilber Lourmat™
311100431 |
400 mW/cm2 | 4 W | N/A | |||||
11870380
|
Vilber Lourmat™
3111-0063-1 |
700 mW/cm2 | 6 W |
564.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
11830430
|
Vilber Lourmat™
3111-0061-1 |
710 mW/cm2 | 6 W |
564.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||
11800250
|
Vilber Lourmat™
3111 0153 1 |
1100 mW/cm2 | 15 W | N/A | |||||
11796658
|
Vilber Lourmat™
3111-2155-1 |
1350/1100 mW/cm2 | 30 W |
969.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||
11766668
|
Vilber Lourmat™
3111-2151-1 |
1780 mW/cm2 | 30 W |
943.00 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
11746668
|
Vilber Lourmat™
3111-2153-1 |
2300 mW/cm2 | 30 W |
943.00 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
Description
- Single wavelength UV lamp with filter
- 6 W UV lamp
- Rapid visualization of CCM plates requiring UV revelation
- Complete with UV tube and filter
- Research and enumeration of E. coli
- UV lamp 254nm
Specifications
1 Ea. | |
UV Lamp |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title