Learn More
Fisherbrand™ UV Lamp, Dual Wavelength
Used as a method of purification. Fisher Scientific™ UV Lamp, Dual Wavelength can be used in combination with other methods to reduce microbial reproduction in purified water.
Brand:  Fisherbrand™ RAUV357B1
 View more versions of this product
Product Code. 15469689
  • 371.00 € / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Accu100 and Accu500 Ultrapure Water Systems
  • Prevents microbial reproduction
Specifications
Specifications
185/254nm
Accu500