Learn More
Faster™ UV Lamp for SafeFAST Safety Cabinets
Magnetic UV lamp for use with SafeFAST Cabinets.
Brand:  Faster™ FX0002154600
Product Code. 15553376
  • 518.00 € / Each
Explore more special offers
Description
Description
  • Plug and play
  • Can be retrofitted
  • Can be positioned and moved around the back wall to ensure maximum exposure to the worksurface
  • Can be removed from the cabinet and attached to the stand if required
Specifications
Specifications
5 to 6 ft. Cabinets
UV
SafeFAST Elite, Classic, Premium, CytoFAST Elite 115, 118, 215, 218