Learn More
Fisherbrand™ UV Sterilization Cabinet with Timer
Limits PCR reaction contamination. Fisherbrand™ UV Sterilization Cabinet with Timer provides a convenient area for setting up PCR reactions in a nucleic acid-free environment.
Brand:  Fisherbrand™ F-UVCAB
 View more versions of this product
Product Code. 15562496
  • 1800.00 € / Each
Explore more special offers
Description
Description
  • UV lights on the cabinets denature nucleic acids in 5 to 30 minutes making them unsuitable for amplification.
  • Incorporates safety features to prevent user exposure to UV light.
  • The UV lights are timer controlled
  • Safety switches on the cabinets doors turn off the UV lights when opened.
  • Shields beta radioactive emissions
  • Specifications
    Specifications
    4 UV Lights, 1 White
    770 mm
    5 x 15 W
    560 mm
    420 mm
    No tray
    19 kg