Learn More
Fisherbrand™ UV Sterilization Cabinet with Timer
Limits PCR reaction contamination. Fisherbrand™ UV Sterilization Cabinet with Timer provides a convenient area for setting up PCR reactions in a nucleic acid-free environment.
Brand:  Fisherbrand™ F-UVCABTY4
2025.12 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 15592496
  • 2600.00 € / Each
Explore more special offers
Description
Description
  • UV lights on the cabinets denature nucleic acids in 5 to 30 minutes making them unsuitable for amplification.
  • Incorporates safety features to prevent user exposure to UV light.
  • The UV lights are timer controlled
  • Safety switches on the cabinets doors turn off the UV lights when opened.
  • Shields beta radioactive emissions
  • Specifications
    Specifications
    4 UV Lights, 1 White
    770 mm
    5 x 15 W
    560 mm
    420 mm
    Tray
    19 kg
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    One moment while we fetch your results.
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Fisherbrand™ UV Sterilization Cabinet with Timer >