missing translation for 'onlineSavingsMsg'
Learn More

Vilber Lourmat™ UV Tables

This ideal, long life UV filter with high transmission power, requires little warm-up and features low noise level, heating and power consumption.

1040.40 €

Specifications

Filtration Area (Metric) 150 x 150 mm
Intensity 10,000 mW/cm2
View More Specs

Products 1
Product Code Brand Filtration Area (Metric) Intensity Price Quantity & Availability  
Product Code Brand Filtration Area (Metric) Intensity Price Quantity & Availability  
11736259
View accessory products View Documents
Vilber Lourmat™
2131-1502-1
150 x 150 mm 10,000 mW/cm2 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • UV fluorescent table
  • Stable, flicker-free light
  • 2-position intensity selector
  • Tilting UV screen
  • UV protection by tilting screen
  • Long life UV filter with high transmission power
  • Stainless steel anti-corrosion cover for easy maintenance
  • 2-position intensity selector: 100%: reading position, 70%: cutting position to minimize the risk of photolysis
  • 25 kHz high frequency electronic ballast for stable flicker-free light
  • Low noise level, heating and power consumption
  • Power supply 100-240 V, 50/60 Hz
  • Compact UV table 312nm, 8 W,  21 x 26cm
Specifications

Specifications

150 x 150 mm
10,000 mW/cm2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.