missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ UV Transilluminator, Benchtop
GREENER_CHOICE

Syngene UV Transilluminator for illumination of any sample excited by UV light is sealed to allow wet gels on surfaces

Brand:  Syngene™ BT-2020M-E60

Product Code. 15829922

  • 1080.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Fixed
241 mm
315 mm
Benchtop use
200 x 200 mm
230 V 60 Hz 230 w
Syngene 2020M-E60
60 Hz
302 nm
80 mm
UV Transilluminator
230 w
315 x 241x 80mm
200 x 200 mm
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers