Learn More
Fisherbrand™ UV Trays
Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems.
Brand:  Fisherbrand™ MS7-UV10
 View more versions of this product
Product Code. 11837573
  • 55.45 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems
Specifications
Specifications
Sub-Gel Midi Horizontal Gel Electrophoresis System
UV Tray
70mm x 100mm