missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vilber Lourmat™ UV Tube
Rich, robust UV (ultraviolet) tube that is ideal for use in many laboratory, research, scientific, and other environments
Brand: Vilber Lourmat™ 9075-2083-1
This item is not returnable.
View return policy
Description
- Standard diameter 16 mm
Specifications
8 W | |
16 mm | |
UV Tube |
365 nm | |
288 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction