missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Uvette™ Routine Pack

Use this cuvette for superb purity, convenience and precision for photometric analysis. Eppendorf™ Uvette™ Routine Packs are disposable and feature a light transmission of 220 nm to 1,600 nm. Deep-lying optical window prevents scratches.

Brand:  Eppendorf™ 0030106318

143.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10145022

  • 151.60 € / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use this cuvette for superb purity, convenience and precision for photometric analysis. Eppendorf™ Uvette™ Routine Packs are disposable and feature a light transmission of 220 nm to 1,600 nm. Deep-lying optical window prevents scratches.

The unique shape of the UVette combines two optical light path lengths to offer greater flexibility. UVettes in routine packs are ideal for routine applications. These packs allow a safe storage of the UVettes in a re-closable box, while offering convenient access to each cuvette for easy filling.
UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10mm and 2mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cuvette Pack
UV-transparent Plastic
2 mm or 10 mm
Biophotometers and Spectrophotometers
50 to 2000 μL
200 Pack
220 to 1600 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Uvette™ Routine Pack > Wavelength: 220 to 1600nm; Qty: 200 Pack

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.