Learn More
Eppendorf™ UVette™
Use these cuvettes for measuring small volumes (≥50μL). The Eppendorf™ UVette™ is a disposable UV/Vis cuvette with a light transmission of 220 to 1,600nm. UVettes offer superb purity, convenience and precision for photometric analysis.
Brand:  Eppendorf™ 0030106300
Product Code. 10412121
Description
Description
Use these cuvettes for measuring small volumes (≥50μL). The Eppendorf™ UVette™ is a disposable UV/Vis cuvette with a light transmission of 220 to 1,600nm. UVettes offer superb purity, convenience and precision for photometric analysis.
  • UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
  • Two built-in optical path lengths in a single cuvette: just turn the UVette 90° to change from 10mm and 2mm
  • Volume markings for 500 and 1,000μL
  • Individually packaged and certified PCR clean and protein-free quality for sensitive (e.g. RNA) and valuable samples
  • Deep-lying optical window prevents scratches
  • Tapered cuvette base for optimal filling, frosted grip for labeling
  • Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers
  • Routine pack for convenient access to each UVette and safe storage in a re-closable box
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cuvette
UV-transparent Plastic
2 mm or 10 mm
Biophotometers and Spectrophotometers
50 to 2000 μL
80 Pack
220 to 1600 nm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products