missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 1 Lady Safety Shoe S2
Sporty, multifunctional safety shoe
Brand: Uvex™ 8562837
123.25 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
ESD certified, EN ISO 20345: 2011 S2 SRC | |
Ladies safety shoe | |
11 mm |
Gray, Pink | |
37 |