missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 1 Safety Shoe S2
Sporty, multifunctional safety shoe
Brand: Uvex™ 8544838
123.25 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Specifications
ESD certified, EN ISO 20345: 2011 S2 SRC | |
Loafer | |
11 mm |
Black | |
38 |