missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ C500 Wet Plus Cut Protection Glove
Click to view available options
Size:
7
9
Description
Specifications
Specifications
Certifications/Compliance | EN 388:2016 (4 X 4 2 C), 407 (X 1 X X X X) |
Coated Area | Back of the hand partially coated |
Color | Lime Green, Anthracite |
Material | HPPE, Polyamide |
Size | 7 |
For Use With (Application) | Automotive industry, glass industry, maintenance/servicing, metal work industry, assembly work, and transport industry |
Glove Cuff Style | Knit |
Model | C500 WET PLUS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction