missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ C500 Wet Plus Cut Protection Glove

Product Code. 15377052
Click to view available options
Size:
7
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for wet areas and oily working conditions.

  • The use of Bamboo TwinFlex® Technology ensures a high level of cut resistance, an exceptional abrasion resistance and a supreme wearing comfort
  • The innovative SoftGrip coating guarantees an excellent dexterity, flexibility and secure grip
  • The uvex C500 wet plus model is certified in accordance with EN 407 for contact heat up to 100 °C
  • Very high cut protection (cut 5)
  • Silicon-free according to imprint test
  • Certified according to Öko-Tex Standard 100
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance EN 388:2016 (4 X 4 2 C), 407 (X 1 X X X X)
    Coated Area Back of the hand partially coated
    Color Lime Green, Anthracite
    Material HPPE, Polyamide
    Size 9
    For Use With (Application) Automotive industry, glass industry, maintenance/servicing, metal work industry, assembly work, and transport industry
    Glove Cuff Style Knit
    Model C500 WET PLUS
    missing translation for 'provideContentCorrection'

    missing translation for 'productTitle'