missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Pheos B-WR Safety Helmet

Helmets are resistant to hot molten metal splash and feature a comfortable, secure fit with continuous width adjustment.

Brand:  Uvex™ 9772530

Product Code. 15336222

  • 16.01 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Resistant to hot molten metal splash and temperatures down to -30°C (EN 367 additional requirement MM)
  • Side euroslot adapters (30 mm) are suitable for the addition of earmuffs or the uvex pheos visor system
  • Suspension harness with wheel ratchet allows continuous width adjustment, for a perfect fit at all times
  • Six-point textile band attachment ensures optimal fit and comfort
  • Available in a wide variety of color options
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EN 397:2012
Blue
Ventilated
Polyethylene with vents
Pheos B-WR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers