missing translation for 'onlineSavingsMsg'
Learn More

UVP Battery 6V

6V Battery for use in Portable UVP UV lamps

Brand:  UVP 45-0005-01

Product Code. 11805883

  • 26.60 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Rechargeable Battery
UVP Lamp
9.5 x 2.8 x 9.4 in. (241 x 71 x 239 mm)
241 mm
71 mm
239 mm
1.8 kg
Field use
Mini built-in flashlight
9.5 in.
2.8 in.
9.4 in.
4 lb.
6 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.