missing translation for 'onlineSavingsMsg'
Learn More

UVP High-Performance Mineralight™ Display Lamps: Replacement Tubes

UVP 25-watt tubes providing hours of UV illumination when used with High-Performance Mineralight Display Lamps.

Brand:  UVP 34-0073-01

 View more versions of this product

Product Code. 11847104

  • 62.90 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 25-watt UV tubes for bright illumination
  • Available with 254nm, 302nm or 365nm UV
  • Use the brackets included with the lamp to attach the lamp to the ceiling or other structure for overhead UV illumination
  • Dimensions: 18.5L x 4.75W x 5 in.H

Fluorescence, Sterilization, Inspection

Specifications

Specifications

Replacement Shortwave Tube
254 nm
Model UVLS-225D
434.84 mm
25 W
Fluorescence, sterilization, inspection
17.12 in.
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.