missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP Lamp Stands
Allows easy and comfortable hands free operation of UV lamps
42.45 € - 197.00 €
Specifications
Product Type | Lamp Stand |
---|---|
For Use With (Equipment) | EL-series UV lamp |
Packaging Quantity | 1 Ea. |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
12611732
|
UVP
18-0063-01 |
J-138 |
197.00 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
11786812
|
UVP
98-0016-03 |
J-129 |
141.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11718231
|
UVP
98-0020-03 |
J-124 |
42.45 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
Description
Allows easy and comfortable hands free operation of UV lamps
- Housing: Varies from metal housing with scratch resistant powder coat to plastic housing
- Allows hands free operation of the lamp
Specifications
Lamp Stand | |
EL-series UV lamp | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title