Learn More
UVP Mini Ultraviolet Lamp Replacement Tube
Replacement tubes for lightweight, compact and financial UV Lamps
Brand:  UVP 34-0004-01
 View more versions of this product
Product Code. 11788231
  • 50.55 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
4 W
Model UVGL-15 and UVSL-14P compact and mini handheld UV lamp
134.9 mm
365/312 nm
5.31 in.
1 Ea.