Learn More
UVP MRL-58 Multiple Ray Lamp Tube
Replacement tube for benchtop lamp
Brand:  UVP 34-0031-01
Product Code. 12812989
  • 50.55 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • UV tube
  • 8w bulb
  • 365nm
Applications:
Gel electrophoresis, Nucleic acid visualization, DNA analysis, Biochemical testing
Specifications
Specifications
8 W
MRL-58 multiple ray lamp and Visi-Blue™ transilluminator
287.7 mm
365 nm
11.33 in.
1 Ea.