Learn More
UVP Replacement UV Light Tubes
Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators
Brand:  UVP 34-0009-02
 View more versions of this product
Product Code. 12883918
  • 27.40 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
15 W
365 nm
Model XX-15L UV bench lamp, ChromaDoc-It™ imaging system and Chromato-Vue™ C-70G, C-71, C-75 large UV viewing cabinets
1 Ea.