Learn More
UVP PhotoDoc-It™ Imaging Systems
Capture, print and save color images for basic gel documentation with this compact, economical laboratory system. UVP™ PhotoDoc-It™ Imaging Systems provide a low cost imaging solution without the need for film.
Brand:  UVP 97-0274-08
 View more versions of this product
Product Code. 11778356
  • 4455.00 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The digital color camera has a 10 mega pixel resolution - the user can view images directly on the camera or the (optional) color LCD screen.
  • The lightweight metal hood provides a compact darkroom environment.
  • Images are saved to the camera memory card or instantly printed as 4x6 in. low cost, color prints using the budget photo printer included.
  • An Ethidium™ Bromide filter for fluorescent stained gels (580 to 630nm) is included inside the hood and can be changed for other filter types.
  • A combination lock secures the camera to the darkroom. LCD shown in picture is an optional accessory. Dimensions: 337mm x 241mm x 432mm The PhotoDoc-It system is a very low cost solution for capturing, printing and saving color images for basic gel documentation without the need for film.
Specifications
Specifications
230V
EtBr Red
432mm
337mm
302-365nm
Yes
Gel Imaging, Colorimetric Imaging, Fluorescence
241mm
16 MP
302-365nm
LM-20
1Pack
DigiCam 70
Imaging System
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products