Learn More
UVP C-15G Mini Chromato-Vue™ UV Viewing Cabinet
Use these cabinets for educational uses, viewing minerals and other applications. UVP™ C-15G Mini Chromato-Vue™ UV Viewing Cabinets provide a darkroom environment with UV fluorescence for viewing materials.
Brand:  UVP 95-0072-07
Product Code. 12823838
  • 728.00 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed with a metal base and plastic top
  • Includes a UVGL-58 UV lamp
  • The bottom panel of the cabinet is removable
  • The lamp fits snugly on top of the cabinet
  • A contrast control filter absorbs UV energy and protects the eyes
  • Soft rubber viewport provides a comfortable viewing area with space for eyeglasses
  • Lightweight access curtain blocks out external light from the cabinet interior
  • Lamps for other mini cabinets are ordered separately
Specifications
Specifications
230V
305mm
C-15G
Mineralogy, Lab Fluorescence, Sterilization, Field Investigation
229mm
267mm
1Pack
UV Cabinet