Learn More
UVP UV Viewing Cabinets
Use these viewing cabinets for a variety of applications such as non-destructive testing, document verification and thin layer chromatography. UVP™ UV Viewing Cabinets feature a removable bottom panel so the cabinet may be placed over large samples or a transilluminator.
Brand:  UVP 95-0257-02
 View more versions of this product
Product Code. 11788201
  • 2390.00 € / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Soft rubber viewport includes a contrast control filter.
  • Control switches are conveniently located for selecting either of the UV wavelengths or the interior overhead white light
  • Easy access to the cabinet interior is via the curtain which also blocks ambient light
  • The C-70G Chromato-Vue Cabinet includes two each of 15-watt shortwave and longwave UV tubes mounted on one side of the cabinet
  • Model C-71 is similar to the C-70G, but has four 15-watt, longwave UV tubes on one side of the unit
  • The 254nm and 365nm UV tubes in the C-75 are positioned on two sides of the cabinet providing uniform UV illumination for fluorescence studies
  • Manufactured from lightweight plastic
Specifications
Specifications
230V
400mm
C-65
Thin Layer Chromatography, Non Destructive Testing, Inspection, Mineralogy, Fluorescence
409mm
493mm
1Pack
UV Chamber