Learn More
Cole-Parmer™ V Series Stuart™ Vortex Mixer
This V Series Stuart™ Vortex Mixer is a compact unit perfect for individual work stations.
328.00 € - 682.00 €
Product Code | Brand | Speed Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Speed Range | Price | Quantity & Availability | |||||
12017134
|
Cole-Parmer™
04729-01 |
Fixed Speed |
328.00 €
Each |
In Stock
Log in to see stock. |
|||||
12027134
|
Cole-Parmer™
04729-03 |
200 to 2500 rpm |
682.00 €
Each |
In Stock
Log in to see stock. |
|||||
15401435
|
Cole-Parmer™
04729-13 |
2500 to 4500 rpm |
539.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- Mix a wide variety of tube sizes and shapes with the included rubber cup head.
- Heavy die-cast body won't dance across your bench.
- Automatic start when the soft rubber cup is pressed down.
- The Vortex mixer provides rapid mixing of samples contained in test-tubes, small flasks and bottles.
- Starts automatically when the rubber cup is depressed and stops once vessel is removed.
- Control the mixing speed by selecting from 200 rpm for more gentle mixing to 2500 rpm for vigorous agitation.
- Heavy die-cast body for stability during use.
- Includes integral retort rod for securing vessels above the vortex action for long-term mixing.